missing translation for 'onlineSavingsMsg'
Learn More

3M™ Reusable Earplugs

The Earplugs 1200 Series are reusable and designed for insertion into the ear canal to help reduce exposure to hazardous levels of noise up to 26 dB.

Brand:  3M™ 7100288287

275.34 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 17873256

  • £413.00 / 300 pairs
Estimated Shipment: 06-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Get comfortable and durable hearing protection with 3M™ Earplugs 1200 Series.
  • These reusable earplugs reduce noise levels up to 26 dB and are fully tested and approved against EN 352-2:2020.
  • They are made from a soft durable Thermoplastic Elastomer (TPE).
  • The specially designed and triple flange shape is tapered to fit most ear canal sizes, offering a comfortable secure fit.
  • The finger grip design keeps insertion and removal simple, convenient and hygienic.
  • Packed in a reusable recyclable cardboard pillow pack, helping keep your earplugs hygienic and together, preventing loss.
  • A reusable Polycon plastic box is available separately.
  • Allowing you the option of a more robust storage solution, but only buying what you need and potentially reducing plastic wastage.
  • 3M™ Earplugs 1271 are available as probed versions for use with the 3M™ E-A-RFit™ validation system.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Regular
26 dB
Flanged
No
No
1271
Corded
Reusable
No
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Brochures

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3M™ Reusable Earplugs > Corded

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.