missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Reusable Earplugs
The Earplugs 1200 Series are reusable and designed for insertion into the ear canal to help reduce exposure to hazardous levels of noise up to 26 dB.
£413.00
Specifications
Size | Regular |
---|---|
Noise Reduction Rating (NRR) | 26 dB |
Plug Shape | Flanged |
Disposable | Reusable |
Metal Detectable | No |
Product Code | Brand | Corded | Model | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Corded | Model | Price | Quantity & Availability | |||||
17883256
|
3M™
7100288286 |
Uncorded | 1261 | N/A | ||||||
17873256
|
3M™
7100288287 |
Corded | 1271 |
£413.00
300 pairs |
Estimated Shipment: 06-05-2025
Log in to see stock. |
|||||
Description
- Get comfortable and durable hearing protection with 3M™ Earplugs 1200 Series.
- These reusable earplugs reduce noise levels up to 26 dB and are fully tested and approved against EN 352-2:2020.
- They are made from a soft durable Thermoplastic Elastomer (TPE).
- The specially designed and triple flange shape is tapered to fit most ear canal sizes, offering a comfortable secure fit.
- The finger grip design keeps insertion and removal simple, convenient and hygienic.
- Packed in a reusable recyclable cardboard pillow pack, helping keep your earplugs hygienic and together, preventing loss.
- A reusable Polycon plastic box is available separately.
- Allowing you the option of a more robust storage solution, but only buying what you need and potentially reducing plastic wastage.
- 3M™ Earplugs are available as probed versions for use with the 3M™ E-A-RFit™ validation system.
Specifications
Regular | |
Flanged | |
No | |
No |
26 dB | |
Reusable | |
No |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
3M™ Reusable Earplugs