missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Reusable Half Face Mask Respirator
3M™ Reusable Half Mask has been designed with user comfort and convenience in mind. Compatible with DIN threaded, 3M™ Filters DT-Series, the HF-301 offers protection against a wide range of respiratory hazards.
£394.00 - £465.00
Specifications
Product Type | Half Facepiece Respirator |
---|---|
For Use With (Equipment) | DIN Threaded, 3M™ Filters DT-Series |
Quantity | 10/Cs. |
Product Code | Brand | Size | Model | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Model | Price | Quantity & Availability | ||||
17766983
|
3M™
7100265024 |
Small | HF-301 |
£465.00
Pack of 10 |
Estimated Shipment: 09-09-2024
Log in to see stock. |
||||
17717003
|
3M™
7100265524 |
Medium | HF-302 |
£394.00
Pack of 10 |
Estimated Shipment: 09-09-2024
Log in to see stock. |
||||
17746993
|
3M™
7100265229 |
Large | HF-303 |
£394.00
Pack of 10 |
Estimated Shipment: 09-09-2024
Log in to see stock. |
||||
Description
- Hybrid reflex seal contours and flexes to the user's face to aid wearer comfort
- Seal check mechanism - when pressed during exhale, the mask inflates slightly showing that a good seal has been achieved
- Integrated voice panel for enhanced voice intelligibility while wearing the mask
- Nose pinch allows for improved integration with eyewear
- Comfortable, durable, low profile head cradle & thick elasticated straps with simple rear fastening
Specifications
Half Facepiece Respirator | |
DIN Threaded, 3M™ Filters DT-Series | |
10/Cs. |