Learn More
3M™ Reusable Low Maintenance Half Face Mask Respirator
Reusable Half Face Masks 6000 Series are made from a lightweight, comfortable elastomeric material with a bayonet-style twin-filter design, cradle head harness and easy-fasten neck strap.
Brand:  3M™ 7100049927
148.05 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 17873246
  • £229.00 / Pack of 8
Estimated Shipment: 22-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For economical, low-maintenance reusable masks that are easy to handle, use 3M™ Reusable Half Face Mask 6000 Series.
  • Our reusable half masks are designed for simplicity, with an elastomeric material that’s lightweight and comfortable to wear over long shifts, a cradle head harness and easy-fasten neck strap.
  • They provide protection against gases, vapours and particulates, and feature a bayonet-style connection that fits to a broad range of twin, lightweight filters, depending on your individual needs.
  • The twin-filter design evenly balances weight across the mask, and the low profile means minimal impact to your field of vision and better compatibility with welding and grinding shields.
  • They can be used with 3M™ Supplied Air Systems for your increased convenience and flexibility, and connect with all 3M™ 2000, 5000 and 6000 Series filters (except 3M™ 6098 or 6099).
  • The 6000 Series has a specially designed range of spares and accessories to help you maintain the right level of comfort and protection when parts become worn or damaged.
  • They’re available in three sizes; small, medium and large.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Gray
Reusable
Thermoplastic Elastomer
Medium
Yes
Bayonet Style
Combination
Reusable Half Facepiece
No
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Brochures
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
3M™ Reusable Low Maintenance Half Face Mask Respirator > Size: Medium