Learn More
Thermo Scientific™ RheFlex™ Two-Piece PEEK Fittings
Thermo Scientific™ RheFlex™ Two-Piece PEEK Fittings provide inert, metal-free connections.
Brand: Thermo Scientific™ 6000-055
This item is not returnable.
View return policy
Description
Thermo Scientific™ RheFlex™ Two-Piece PEEK Fittings provide inert, metal-free connections.
- Slotted back-side of the ferrule is squeezed down onto the tube by the mating conical surface of the nut
- May be used on 1/16 in. metal or plastic tubing reliably up to 5000 psi
- Reusable ferrule and nut
Specifications
Fitting Set, Short | |
1/16 in. Short fitting set; 5/Pk. | |
PEEK Tubing | |
PEEK |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.