Learn More
Thermo Scientific™ RheFlex™ Two-Piece PEEK Fittings
Thermo Scientific™ RheFlex™ Two-Piece PEEK Fittings provide inert, metal-free connections.
£28.85 - £41.46
Specifications
For Use With (Equipment) PEEK Tubing
Material PEEK
View More Specs
Products 4
Product Code Brand Type Price Quantity & Availability  
Product Code Brand Type Price Quantity & Availability  
11909234
View Documents
Thermo Scientific™
6000-051
Replacement Ferrule This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11919234
View Documents
Thermo Scientific™
6000-054
Fitting Set, Standard Length This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10669865
View Documents
Thermo Scientific™
6000-055
Fitting Set, Short This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11959214
View Documents
Thermo Scientific™
6000-066
Fitting Set, X-Long This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
Thermo Scientific™ RheFlex™ Two-Piece PEEK Fittings provide inert, metal-free connections.
  • Slotted back-side of the ferrule is squeezed down onto the tube by the mating conical surface of the nut
  • May be used on 1/16 in. metal or plastic tubing reliably up to 5000 psi
  • Reusable ferrule and nut
Specifications
Specifications
PEEK Tubing
PEEK
Videos
SDS
Documents
Documents
Product Certifications
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.