missing translation for 'onlineSavingsMsg'
Learn More
Rhenium powder, -22 mesh, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals Product Code.: 11310909

Rhenium powder, -22 mesh, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals

Product Code. 11310909
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11310909

Brand: Thermo Scientific Alfa Aesar 010673.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-15-5
Molecular Formula Re
Molecular Weight (g/mol) 186.21
MDL Number MFCD00011195
InChI Key WUAPFZMCVAUBPE-UHFFFAOYSA-N
Synonym rhenium, elementar, renio, unii-7yhu292iny, 7yhu292iny, foil, ion re7, rhenium, ion re7, atom, hydride, pellets
PubChem CID 23947
ChEBI CHEBI:49882
IUPAC Name rhenium
SMILES [Re]

Specifications

UN Number UN3089
Quantity 1 g
Percent Purity 99.999%
Grade Puratronic™
Mesh Size −22 Mesh
Form Powder
Assay (metals basis)
Chemical Name or Material Rhenium powder

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX22F008 Certificate TypeCertificate of Analysis Date20/12/2024 Catalog Number010673.03,010673.06
Lot NumberM13D010 Certificate TypeCertificate of Analysis Date22/08/2024 Catalog Number010673.03,010673.06
Lot NumberM04K032 Certificate TypeCertificate of Analysis Date17/01/2024 Catalog Number010673.03,010673.06
Lot NumberX19I051 Certificate TypeCertificate of Analysis Date04/11/2022 Catalog Number010673.03,010682.14,010673.06
Lot NumberX22F008 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number010673.03,010682.14,010673.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Rhenium powder, -22 mesh, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.