missing translation for 'onlineSavingsMsg'
Learn More
Rhenium powder, 99.99%, Thermo Scientific Chemicals Product Code.: 11321418

Rhenium powder, 99.99%, Thermo Scientific Chemicals

Product Code. 11321418
2 g
Click to view available options
Quantity:
0.2 g
2 g
10 g
50 g
This item is not returnable. View return policy

Product Code. 11321418

Brand: Thermo Scientific Alfa Aesar 010313.04

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-15-5
Molecular Formula Re
Molecular Weight (g/mol) 186.21
MDL Number MFCD00011195
InChI Key WUAPFZMCVAUBPE-UHFFFAOYSA-N
PubChem CID 23947
ChEBI CHEBI:49882
IUPAC Name rhenium
SMILES [Re]

Specifications

Appearance Powder
Assay Percent Range 99.99% (Metals basis)
Quantity 2 g
Synonym rhenium, elementar, renio, unii-7yhu292iny, 7yhu292iny, foil, ion re7, rhenium, ion re7, atom, hydride, pellets
Mesh Size -325 Mesh
Form Powder
Chemical Name or Material Rhenium

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberR29K001 Certificate TypeCertificate of Analysis Date25/06/2024 Catalog Number010313.SS,010313.18,010313.04
Lot NumberY08I033 Certificate TypeCertificate of Analysis Date18/11/2022 Catalog Number010313.SS,010313.18,010313.04,010313.09
Lot NumberM26I018 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number010313.SS,010313.18,010313.04,010313.09
Lot NumberQ14G001 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number010313.SS,010313.18,010313.04,010313.09
Lot NumberK20T018 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number010313.SS,010313.18,010313.04,010313.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Rhenium powder, 99.99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.