Learn More
Rhenium(VII) oxide, 99.995% (metals basis), Thermo Scientific Chemicals
£121.61 - £335.00
Chemical Identifiers
CAS | 1314-68-7 |
---|---|
Molecular Formula | O7Re2 |
Molecular Weight (g/mol) | 484.407 |
MDL Number | MFCD00011199 |
InChI Key | NBGOSNNAAHDRLK-UHFFFAOYSA-N |
Synonym | rhenium heptoxide, rhenium vii oxide, re2o7, dirhenium heptoxide, dirhenium heptaoxide, rhenium oxide re2o7, trioxo-trioxorheniooxy rhenium, trioxo trioxorheniooxy rhenium, rhenium vii oxide-re, rhenium vii oxide trace metals basisShow More |
PubChem CID | 123106 |
IUPAC Name | trioxo(trioxorheniooxy)rhenium |
SMILES | O=[Re](=O)(=O)O[Re](=O)(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15412347
|
Thermo Scientific Alfa Aesar
045538.04 |
2 g |
£121.61
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15422347
|
Thermo Scientific Alfa Aesar
045538.09 |
10 g | N/A | ||||||
Description
Rhenium(VII) oxide is a precursor to organorhenium oxides which catalyze olefin oxidation, metathesis and other transformations. It is also used as a catalyst for hemiacetal substitution and as an oxidant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1314-68-7 | |
484.407 | |
NBGOSNNAAHDRLK-UHFFFAOYSA-N | |
123106 | |
O=[Re](=O)(=O)O[Re](=O)(=O)=O |
O7Re2 | |
MFCD00011199 | |
rhenium heptoxide, rhenium vii oxide, re2o7, dirhenium heptoxide, dirhenium heptaoxide, rhenium oxide re2o7, trioxo-trioxorheniooxy rhenium, trioxo trioxorheniooxy rhenium, rhenium vii oxide-re, rhenium vii oxide trace metals basisShow More | |
trioxo(trioxorheniooxy)rhenium |
Specifications
360°C | |
1314-68-7 | |
Crystalline | |
MFCD00011199 | |
Soluble in water and alcohol. | |
O=[Re](=O)(=O)O[Re](=O)(=O)=O | |
484.407 | |
484.4 | |
Moisture sensitive | |
Rhenium(VII) oxide |
297°C | |
Yellow | |
O7Re2 | |
rhenium heptoxide, rhenium vii oxide, re2o7, dirhenium heptoxide, dirhenium heptaoxide, rhenium oxide re2o7, trioxo-trioxorheniooxy rhenium, trioxo trioxorheniooxy rhenium, rhenium vii oxide-re, rhenium vii oxide trace metals basis | |
NBGOSNNAAHDRLK-UHFFFAOYSA-N | |
trioxo(trioxorheniooxy)rhenium | |
123106 | |
Odorless | |
6.103 |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'dotInformation' : DOT Class: 5.1, Packaging Group: III
missing translation for 'einecsNumber' : 215-241-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
- Rhenium(VII) oxide
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Rhenium oxide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.