missing translation for 'onlineSavingsMsg'
Learn More
Rhodium, 5% on carbon, dry, Thermo Scientific Chemicals Product Code.: 11428536

Rhodium, 5% on carbon, dry, Thermo Scientific Chemicals

Product Code. 11428536
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11428536

Brand: Thermo Scientific Alfa Aesar A15965.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rhodium, 5% on carbon is a useful reagent for the conversation of nitriles to primary amines and the hydrogenation of aromatic rings, pyridines, aldehydes and sugars. It is also used for the reduction of oximes to amines and for the reduction of nitrobenzene to phenylhydroxylamine using hydrazine hydrate as the hydrogen source.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Rhodium, 5% on carbon is a useful reagent for the conversation of nitriles to primary amines and the hydrogenation of aromatic rings, pyridines, aldehydes and sugars. It is also used for the reduction of oximes to amines and for the reduction of nitrobenzene to phenylhydroxylamine using hydrazine hydrate as the hydrogen source.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, halogens, chlorine trifluoride and oxygen difluoride.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-16-6
Molecular Formula Rh
Molecular Weight (g/mol) 102.91
MDL Number MFCD00011201 MFCD03458392
InChI Key MHOVAHRLVXNVSD-UHFFFAOYSA-N
Synonym fume, on carbon, rhodium, elemental, rodio, black, rhodium, metal, unii-dmk383dsac, on alumina, rh/al2o3, dmk383dsac
PubChem CID 23948
ChEBI CHEBI:33359
IUPAC Name rhodium
SMILES [Rh]

Specifications

UN Number UN1325
Quantity 1 g
Merck Index 14,8186
Solubility Information Insoluble in water.
Odor Odorless
Chemical Name or Material Rhodium, 5% on carbon, dry

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10251540 Certificate TypeCertificate of Analysis Date07/08/2024 Catalog NumberA15965.03,A15965.14,A15965.06
Lot Number10242371 Certificate TypeCertificate of Analysis Date27/07/2022 Catalog NumberA15965.03,A15965.14,A15965.06
Lot NumberM12481 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15965.03,A15965.14,A15965.06
Lot Number10232076 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15965.03,A15965.14,A15965.06
Lot NumberJA0060 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15965.03,A15965.14,A15965.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Rhodium, 5% on carbon, dry, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.