Learn More
Rhodium(III) chloride hydrate, 38% Rh, Thermo Scientific Chemicals
£865.00 - £3566.00
Chemical Identifiers
CAS | 20765-98-4 |
---|---|
Molecular Formula | Cl3Rh |
Molecular Weight (g/mol) | 209.26 |
MDL Number | MFCD00149839 |
InChI Key | SONJTKJMTWTJCT-UHFFFAOYSA-K |
IUPAC Name | rhodium(3+) trichloride |
SMILES | [Cl-].[Cl-].[Cl-].[Rh+3] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10561641
|
Thermo Scientific Acros
195520010 |
1 g | Glass bottle |
£865.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10571641
|
Thermo Scientific Acros
195520050 |
5 g | Glass bottle |
£3566.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
20765-98-4 | |
209.26 | |
SONJTKJMTWTJCT-UHFFFAOYSA-K | |
[Cl-].[Cl-].[Cl-].[Rh+3] |
Cl3Rh | |
MFCD00149839 | |
rhodium(3+) trichloride |
Specifications
100.0°C | |
Brown to Brown-Red | |
Cl3Rh | |
MFCD00149839 | |
Solubility in water: soluble. Other solubilities: soluble in alchol and hcl, insoluble in ether | |
[Cl-].[Cl-].[Cl-].[Rh+3] | |
209.26 | |
38% (Rh) |
20765-98-4 | |
Crystalline Powder | |
RhCl3·xH2O | |
10,343; 11,460; 14,271; 16,292; 17,366 | |
SONJTKJMTWTJCT-UHFFFAOYSA-K | |
rhodium(3+) trichloride | |
209.26 | |
Rhodium(III) chloride hydrate |
Safety and Handling
GHS H Statement
Causes serious eye damage.
Harmful if swallowed.
Suspected of causing genetic defects.
May be corrosive to metals.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
GHS Signal Word: Danger
- Rhodium(III) chloride hydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Germ cell mutagenicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
Hazard Statement
- H318-Causes serious eye damage.
- H341-Suspected of causing genetic defects.
- H302-Harmful if swallowed.
- H290-May be corrosive to metals.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.,IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/physician.,Immediately call a POISON CENTER/doctor/physician.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.,Wear protective gloves/protective clothing/eye protection/face protection.
RUO – Research Use Only