missing translation for 'onlineSavingsMsg'
Learn More
Rhodium wire, 0.25mm (0.01in) dia, 99.8% (metals basis), Thermo Scientific Chemicals Product Code.: 11317317

Rhodium wire, 0.25mm (0.01in) dia, 99.8% (metals basis), Thermo Scientific Chemicals

Product Code. 11317317
100 cm
Click to view available options
Quantity:
25 cm
100 cm
This item is not returnable. View return policy

Product Code. 11317317

Brand: Thermo Scientific Alfa Aesar 011577.BY

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-16-6
Molecular Formula Rh
Molecular Weight (g/mol) 102.91
MDL Number MFCD00011201 MFCD03458392
InChI Key MHOVAHRLVXNVSD-UHFFFAOYSA-N
Synonym fume, on carbon, rhodium, elemental, rodio, black, rhodium, metal, unii-dmk383dsac, on alumina, rh/al2o3, dmk383dsac
PubChem CID 23948
ChEBI CHEBI:33359
IUPAC Name rhodium
SMILES [Rh]

Specifications

Quantity 100 cm
Percent Purity 99.8%
Weight ≈0.61g/m
Form Wire
Assay (metals basis)
Chemical Name or Material Rhodium wire, 0.25mm (0.01 in.) dia.

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU12I017 Certificate TypeCertificate of Analysis Date18/08/2022 Catalog Number011577.BY,011555.30,011555.14,011555.A1
Lot NumberQ26I014 Certificate TypeCertificate of Analysis Date11/08/2022 Catalog Number011577.BY,011578.CC,011577.BU
Lot NumberZ17H055 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number011577.BY,011555.30,011555.14,011555.A1
Lot NumberA13Y021 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number011577.BY,011578.CC,011577.BU
Lot NumberJ20Q084 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number011577.BY,011578.CC,011577.BU

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Rhodium wire, 0.25mm (0.01in) dia, 99.8% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.