missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rhodizonic acid, disodium salt, 98%, Thermo Scientific Chemicals
£31.80 - £365.00
Chemical Identifiers
CAS | 523-21-7 |
---|---|
Molecular Formula | C6Na2O6 |
Molecular Weight (g/mol) | 214.04 |
MDL Number | MFCD00001650 |
InChI Key | SXWPCWBFJXDMAI-UHFFFAOYSA-L |
Synonym | sodium rhodizonate, sodium 3,4,5,6-tetraoxocyclohex-1-ene-1,2-bis olate, rhodizonic acid disodium salt, unii-9qf0329l3g, rhodizonic acid sodium salt, rhodizonic acid, disodium salt, disodium 3,4,5,6-tetraoxocyclohexene-1,2-diolate, 3,4,5,6-tetraoxocyclohexene-1,2-diol disodium salt, disodium tetraoxocyclohex-1-ene-1,2-bis olate, dipotassium tetraoxocyclohex-1-ene-1,2-bis olate |
PubChem CID | 68225 |
SMILES | [Na+].[Na+].[O-]C1=C([O-])C(=O)C(=O)C(=O)C1=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10428870
|
Thermo Scientific Acros
132340050 |
5 g | Glass bottle | ||||||
11984861
|
Thermo Scientific Acros
132340250 |
25 g | Glass bottle | ||||||
15119183
|
Thermo Scientific Acros
132341000 |
100 g | Glass bottle | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
523-21-7 | |
214.04 | |
SXWPCWBFJXDMAI-UHFFFAOYSA-L | |
68225 |
C6Na2O6 | |
MFCD00001650 | |
sodium rhodizonate, sodium 3,4,5,6-tetraoxocyclohex-1-ene-1,2-bis olate, rhodizonic acid disodium salt, unii-9qf0329l3g, rhodizonic acid sodium salt, rhodizonic acid, disodium salt, disodium 3,4,5,6-tetraoxocyclohexene-1,2-diolate, 3,4,5,6-tetraoxocyclohexene-1,2-diol disodium salt, disodium tetraoxocyclohex-1-ene-1,2-bis olate, dipotassium tetraoxocyclohex-1-ene-1,2-bis olate | |
[Na+].[Na+].[O-]C1=C([O-])C(=O)C(=O)C(=O)C1=O |
Specifications
>300.0°C | |
Gray-Green to Green | |
98% | |
MFCD00001650 | |
15,8799 | |
(0.2% in water) Clear | |
[Na+].[Na+].[O-]C1=C([O-])C(=O)C(=O)C(=O)C1=O | |
214.04 | |
214.04 | |
Authentic | |
Rhodizonic acid, disodium salt |
523-21-7 | |
Fine Crystalline Powder | |
C6Na2O6 | |
08,536 | |
sodium rhodizonate, sodium 3,4,5,6-tetraoxocyclohex-1-ene-1,2-bis olate, rhodizonic acid disodium salt, unii-9qf0329l3g, rhodizonic acid sodium salt, rhodizonic acid, disodium salt, disodium 3,4,5,6-tetraoxocyclohexene-1,2-diolate, 3,4,5,6-tetraoxocyclohexene-1,2-diol disodium salt, disodium tetraoxocyclohex-1-ene-1,2-bis olate, dipotassium tetraoxocyclohex-1-ene-1,2-bis olate | |
SXWPCWBFJXDMAI-UHFFFAOYSA-L | |
disodium;3,4,5,6-tetraoxocyclohexene-1,2-diolate | |
68225 | |
98% | |
(0.2% aq. soln.) clear |
Safety and Handling
missing translation for 'einecsNumber' : 208-340-3
RUO – Research Use Only