missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Rhosin hydrochloride

Rho GTPase inhibitor

£124.00 - £428.00

Specifications

Purity >98%
Product Type Rhosin hydrochloride
Molecular Weight (g/mol) 394.86
Inhibitors Rho GTPase Inhibitors
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18764709
View Documents Promotion Details
Tocris Bioscience™
5003/50
50 mg
£428.00
50mg
Please to purchase this item. Need a web account? Register with us today!
18798718
View Documents
Tocris Bioscience™
5003/10
10 mg
£124.00
10mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Rho GTPase inhibitor (Kd = 354 nM). Inhibits binding of RhoA to guanine nucleotide exchange factors (GEFs) including LARG, DBL, LBC, p115 RhoGEF and PDZ RhoGEF. Has no effect on the binding of Rac1 or Cdc42 to their respective GEFs. Suppresses RhoA/C-mediated breast cancer cell migration and invasion in vitro. Also induces neurite outgrowth in synergy with NGF.
Specifications

Specifications

>98%
Rhosin hydrochloride
394.86
Rho GTPase Inhibitors
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only