missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ RiboLock RNase Inhibitor (40 U/μL)
RiboLock RNase Inhibitor specifically inhibits the activity of RNases A, B, and C, protecting RNA under a variety of reaction conditions.
£50.25 - £1084.00
Specifications
Concentration | 40 U/μL |
---|---|
Compatible Buffer | Storage Buffer |
Product Type | RNase Inhibitor |
Product Line | RiboLock™ |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10859710
|
Thermo Scientific™
EO0381 |
2500 U |
£50.25
2500 units |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11581505
|
Thermo Scientific™
EO0382 |
4 x 2500 U |
£197.00
Pack of 4 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10389109
|
Thermo Scientific™
EO0384 |
24 x 2500 U |
£1084.00
Pack of 24 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
RiboLock RNase Inhibitor inhibits the activity of RNases A , B, and C by binding them in a noncompetitive mode at a 1:1 ratio. It does not inhibit eukaryotic RNases T1, T2, U1, U2, CL3 as well as prokaryotic RNases I and H.
Highlights
- Performs under a wide range of reaction conditions
- Protects RNA from degradation at temperatures up to 55°C
Specifications
40 U/μL | |
Storage Buffer | |
RNase Inhibitor | |
RiboLock™ |
For Research Use Only. Not for use in diagnostic procedures.