Learn More
Epredia™ Richard-Allan Scientific™ Elastic Stain, Van Gieson Stain Solution
Demonstrate the atrophy, thinning and loss of elastic tissue with Epredia™ Richard-Allan Scientific™ Elastic Stain.
Brand: Epredia™ 88000
Code : A0
Additional Details : Weight : 0.01000kg
Description
- Based on the Verhoeff Elastic technique
- Also used for identifying the breaks, splitting and reduplication of elastic lamellae that occur in vascular diseases
- Elastic fibers are stained blue-black to-black with red collagen and a yellow background
Specifications
Sodium Thiosulfate Solution |