Learn More
Epredia™ Richard-Allan Scientific™ Elastic Stain, Van Gieson Stain Solution
Demonstrate the atrophy, thinning and loss of elastic tissue with Epredia™ Richard-Allan Scientific™ Elastic Stain.
£52.95 - £153.00
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
12513637
|
Epredia™
88007 |
Alcoholic Hematoxylin Solution |
£96.95
250mL |
Estimated Shipment: 04-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12543627
|
Epredia™
88000 |
Sodium Thiosulfate Solution |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12533637
|
Epredia™
88009 |
Weigert's Iodine Solution |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12543637
|
Epredia™
88011 |
Van Gieson Stain |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Based on the Verhoeff Elastic technique
- Also used for identifying the breaks, splitting and reduplication of elastic lamellae that occur in vascular diseases
- Elastic fibers are stained blue-black to-black with red collagen and a yellow background