Learn More
Epredia™ Richard-Allan Scientific™ Elastic Stain, Van Gieson Stain Solution
Demonstrate the atrophy, thinning and loss of elastic tissue with Epredia™ Richard-Allan Scientific™ Elastic Stain.
Brand:  Epredia™ 88011
Code : NEW
Additional Details : CAS Number : 88-89-1 Weight : 0.25000kg Transport : UN number : 3265 Chem class : 8 Pack group : III
 View more versions of this product
Product Code. 12543637
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Based on the Verhoeff Elastic technique
  • Also used for identifying the breaks, splitting and reduplication of elastic lamellae that occur in vascular diseases
  • Elastic fibers are stained blue-black to-black with red collagen and a yellow background
Specifications
Specifications
Van Gieson Stain
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Chromaview Van Geison
Signal Word
  • Warning
Hazard Category
  • Specific target organ toxicity Category 3
Hazard Statement
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : diazolidinyl urea, picric acid, fuchsineacid