missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ High-Quality Dehydrants

Reduce spills with easy-to-pour Epredia™ High-Quality Dehydrants.

£83.43

Specifications

Color Colorless
View More Specs

Products 1
Product Code Brand Product Type Concentration Quantity Price Quantity & Availability  
Product Code Brand Product Type Concentration Quantity Price Quantity & Availability  
12510017
View Documents
Epredia™
9511
Alcohol 99% 1 gal. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Reduce spills with easy-to-pour Epredia™ High-Quality Dehydrants.

  • Formulated to rigid specifications, providing laboratories with high-quality products
  • Packaged in high-density polyethylene, which is safer and lighter than glass
Specifications

Specifications

Colorless
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : Hygroscopic

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H371-May cause damage to organs.

Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P235-Keep cool.
  • P314-Get medical advice/attention if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P370+P378-In case of fire: Use CO2, dry chemical, or foam to extinguish.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : isopropyl alcohol

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.