missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ High-Quality Dehydrants

Reduce spills with easy-to-pour Epredia™ High-Quality Dehydrants.

Brand:  Epredia™ 9511


 View more versions of this product

Product Code. 12510017

This item is not returnable. View return policy

Description

Description

Reduce spills with easy-to-pour Epredia™ High-Quality Dehydrants.

  • Formulated to rigid specifications,providing laboratories with high-quality products
  • Packaged in high-density polyethylene, which is safer and lighter than glass
TRUSTED_SUSTAINABILITY
Specifications

Specifications

81° to 83°C
Incompatible Materials: Strong oxidizing agents, Acids, Halogens, Acid anhydrides
99%
Isopropyl Alcohol
Liquid
Colorless
Keep away from heat, flames and sparks. Exposure to moist air or water.
-89.5°C
1 gal., 4/case
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Epredia™ High-Quality Dehydrants > Isopropyl Alcohol; 1 Gal. (3.79L)

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.