missing translation for 'onlineSavingsMsg'
Learn More
Rifabutin, 98%, Thermo Scientific Chemicals Product Code.: 15721769

Rifabutin, 98%, Thermo Scientific Chemicals

Product Code. 15721769
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15721769

Brand: Thermo Scientific Acros 461210010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rifabutin, CAS # 72559-06-09, is a broad-spectrum antibiotic with activity against mycobacteria, gram-positive and gram-negative bacteria.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Rifabutin is a compound with antibacterial activity most commonly used against Mycobacterium tuberculosis
  • Rifabutin is a derivative of Rifamycin S that acts through blocking RNA polymerase subunit beta (RpoB)
  • Rifabutin is an inducer as well as a substrate of CYP3A, a cytochrome P450 enzyme

Applications

  • Rifabutin may be used in pharmacokinetic laboratory studies and high-throughput drug screens
  • May be used in laboratory studies to determine drug resistance mutations present in Mycobacterium tuberculosis strains
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72559-06-9
Molecular Formula C46H62N4O11
Molecular Weight (g/mol) 847.02
MDL Number MFCD00866816
InChI Key ATEBXHFBFRCZMA-VXTBVIBXSA-N
Synonym rifabutin, ansamycin, rifabutine, mycobutin, ansatipine, ansatipin, alfacid, rifabutina, rifabutinum, antibiotic lm 427
PubChem CID 57448257
IUPAC Name (7S,9E,11S,12R,13S,14R,15R,16R,17S,18S,19E,21Z)-2,15,17-trihydroxy-11-methoxy-3,7,12,14,16,18,22-heptamethyl-1'-(2-methylpropyl)-6,23,32-trioxo-8,33-dioxa-24,27,29-triazaspiro[pentacyclo[23.6.1.1⁴,⁷.0⁵,³¹.0²⁶,³⁰]tritriacontane-28,4'-piperidin]-1,3,5(31),9,19,21,25,29-octaen-13-yl acetateShow MoreShow Less
SMILES CO[C@H]1\C=C\O[C@@]2(C)OC3=C(C)C(O)=C4C(=O)C(NC(=O)\C(C)=C/C=C/[C@H](C)[C@H](O)[C@@H](C)[C@@H](O)[C@@H](C)[C@H](OC(C)=O)[C@@H]1C)=C1NC5(CCN(CC(C)C)CC5)N=C1C4=C3C2=OShow MoreShow Less

Specifications

Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 847
Percent Purity 98%
Chemical Name or Material Rifabutin

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467861 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number461210050,461210010
Lot NumberA0455727 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number461210050,461210010
Lot NumberA0451198 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number461210050,461210010
Lot NumberA0397475 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number461210050,461210010
Lot NumberA0399461 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number461210050,461210010

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Rifabutin, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.