Learn More
Quickfit™ Still Head Vertical Recovery Adapter
Used in distillation assemblies in which the condenser is assembled in the vertical mode
Brand: Quickfit SH2/33
28.64 GBP valid until 2024-07-31
Use promo code "21640" to get your promotional price.
Code : Z4
Additional Details : Weight : 0.11000kg
Description
Used in distillation assemblies in which the condenser is assembled in the vertical mode
- Used in distillation assemblies in which the condenser is assembled in the vertical mode
Specifications
Used in distillation assemblies in which the condenser is assembled in the vertical mode | |
Vertical Recovery Still Head | |
Borosilicate Glass | |
ISO 3585 |