Learn More
Quickfit™ Still Head Vertical Recovery Adapter
Used in distillation assemblies in which the condenser is assembled in the vertical mode
£48.75 - £182.00
Specifications
For Use With (Application) | Used in distillation assemblies in which the condenser is assembled in the vertical mode |
---|---|
Type | Vertical Recovery Still Head |
Material | Borosilicate Glass |
Certifications/Compliance | ISO 3585 |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
15507970
|
Quickfit
SH2/22 |
Cone Size: 19/26 |
£182.00
Pack of 5 |
Estimated Shipment: 17-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10438010
|
Quickfit
SH2/33 |
19/26 (Condenser), 19/26 (flask) |
£48.75
Each |
In Stock
Log in to see stock. |
|||||
Description
Used in distillation assemblies in which the condenser is assembled in the vertical mode
- Used in distillation assemblies in which condenser is assembled in the vertical mode
Specifications
Used in distillation assemblies in which the condenser is assembled in the vertical mode | |
Vertical Recovery Still Head | |
Borosilicate Glass | |
ISO 3585 |