missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Rite-On Frosted Thick Slide
Label slides quickly and easily with Epredia™ Gold Seal™ Rite-On™ and Ultra Frost™ Frosted Microscope slides, made from hard, colorless, noncorrosive glass.
£1261.00
Specifications
Material | Glass |
---|---|
Precleaned | Yes |
Width (Metric) | 25 mm |
Color | White |
Quantity | 72 Slides/Box, 50 Boxes/Case |
Product Code | Brand | Corner Style | Frosted | Thickness (Metric) | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Corner Style | Frosted | Thickness (Metric) | Length (Metric) | Price | Quantity & Availability | |||||
15307940
|
Epredia™
K73050 |
90° | White Epoxy Ink Frosted End, 1 Side | 1 mm | 75 mm | N/A | ||||||
Description
All finished slides are sealed with a desiccant in an air tight, lint-free, hermetically sealed Moistur-Barrier™ package, keeping slides dry during shipping and storage. Slides work well in hematology, histology, cytology, microbiology and urinalysis; as well as other laboratory and research applications.- Smooth marking area for effortless writing, automated printing, or printed labels
- Each case features an inner barrier-bag sealed with desiccant packs to protect from moisture contamination
- Slide boxes convert into convenient slide storage files
- Reduced iron content for low self-fluorescence
- Manufactured from pure, water-white glass
- Precleaned for immediate use
- Part No. 3032-002 is used for fluorescent antibody examination
Specifications
Glass | |
25 mm | |
72 Slides/Box, 50 Boxes/Case |
Yes | |
White | |
1 in. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Epredia™ Rite-On Frosted Thick Slide