missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNA Isolation, RA1 Lysis Buffer, Macherey-Nagel™
£70.10 - £154.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11912412
|
Macherey Nagel Bioanalysis
740961 |
50 mL |
£70.10
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10335832
|
Macherey Nagel Bioanalysis
740961.500 |
500 mL | N/A | ||||||
Description
Cells are lysed by incubation in a solution containing chaotropic ions. This lysis buffer inactivates RNases creating appropriate binding conditions favouring adsorption of RNA to the silica membrane. After lysis, homogenisation and reduction of viscosity are achieved by filtration with NucleoSpin™ filter units. Contaminating DNA bound to the membrane is removed by a DNase I solution directly applied to the membrane during preparation. Optimal conditions for DNase are achieved by washing the membrane with a specific desalting buffer before treatment. Salts, metabolites and macromolecular cellular components are removed by simple washing with two different buffers. Total RNA is finally eluted with RNase-free water. The NucleoSpin™ RNA Plant kit features two alternative lysis buffers: in most cases, use of buffer RA1 is recommended for lysis due to its strong denaturing properties. The presence of peculiar metabolites in a variety of plant tissues or fungi may lead to solidification of the lysate, resulting in non-processible slurry. In such cases, the alternative buffer, RAP is the buffer of choice. For thorough homogenisation and reduction of lysate viscosity, NucleoSpin™ filter units are also provided. Kit components: NucleoSpin™ RNA columns with 2mL collecting tubes, 2mL collecting tubes, 1.5mL microcentrifuge tubes, NucleoSpin™ filters, buffers, RNase-free DNase I, DNase I reaction buffer, RNase-free water.- Silica membrane technology
- Yield: 3μg to 70μg from 100μg plant material
- Sample material: 1mg to 100mg tissue
- Elution volume: 60μL
- Fragment size: 200b to 20kb
- Binding capacity: 200μg
- Preparation time: 30min/6 preps
- Format: mini spin column
- DNase I included
- NucleoSpin filter columns included for homogenisation of lysate and reduction of viscosity
- Parallel purification of DNA possible by using the NucleoSpin RNA/DNA buffer set NucleoSpin™ RNA Plant is designed for isolation of DNA-free high quality RNA from a wide variety of plant and fungal samples.
Specifications
Lysis Buffer RA1 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Supplemental information
- Buffer RA1
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H302-Harmful if swallowed.
- H412-Harmful to aquatic life with long lasting effects.
Supplemental information
- MIXTURE LIST-Contains : guanidinium thiocyanate