missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Lysis Buffer RA1
Lysis Buffer RA1 to be used with NucleoSpin RNA, NucleoSpin RNA Midi, NucleoSpin RNA XS, NucleoSpin 8 / 96 RNA, or NucleoSpin RNA Plant kits
Brand: Macherey-Nagel™ 740961.500
This item is not returnable.
View return policy
Description
Cells are lysed by incubation in a solution containing chaotropic ions. This lysis buffer inactivates RNases creating appropriate binding conditions favouring adsorption of RNA to the silica membrane. After lysis, homogenisation and reduction of viscosity are achieved by filtration with NucleoSpin™ filter units. Contaminating DNA bound to the membrane is removed by a DNase I solution directly applied to the membrane during preparation. Optimal conditions for DNase are achieved by washing the membrane with a specific desalting buffer before treatment. Salts, metabolites and macromolecular cellular components are removed by simple washing with two different buffers. Total RNA is finally eluted with RNase-free water. The NucleoSpin™ RNA Plant kit features two alternative lysis buffers: in most cases, use of buffer RA1 is recommended for lysis due to its strong denaturing properties. The presence of peculiar metabolites in a variety of plant tissues or fungi may lead to solidification of the lysate, resulting in non-processible slurry. In such cases, the alternative buffer, RAP is the buffer of choice. For thorough homogenisation and reduction of lysate viscosity, NucleoSpin™ filter units are also provided. Kit components: NucleoSpin™ RNA columns with 2mL collecting tubes, 2mL collecting tubes, 1.5mL microcentrifuge tubes, NucleoSpin™ filters, buffers, RNase-free DNase I, DNase I reaction buffer, RNase-free water.- Silica membrane technology
- Yield: 3μg to 70μg from 100μg plant material
- Sample material: 1mg to 100 mg tissue
- Elution volume: 60μL
- Fragment size: 200b to 20kb
- Binding capacity: 200μg
- Preparation time: 30 min./6 preps
- Format: mini spin column
- DNase I included
- NucleoSpin filter columns included for homogenisation of lysate and reduction of viscosity
- Parallel purification of DNA possible by using the NucleoSpin RNA/DNA buffer set NucleoSpin™ RNA Plant is designed for isolation of DNA-free high quality RNA from a wide variety of plant and fungal samples.
Specifications
Lysis Buffer RA1 | |
RNA isolation | |
500 mL |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Safety and Handling
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Macherey-Nagel™ Lysis Buffer RA1 > Quantity: 500 mL
Spot an opportunity for improvement?Share a Content Correction