Learn More
MP Biomedicals™ RNase-Erase Decontaminants
Completely removes RNase contamination from glass and plastic surfaces
Brand:  MP Biomedicals™ 04821682
136.87 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.
 View more versions of this product
Product Code. 11421722
  • £153.00 / 250mL
Estimated Shipment: 09-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Eliminates the need to bake glassware or use DEPC.
  • Leaves no residues that might inhibit enzymatic reactions
  • Supplied as ready-to-use formula
TRUSTED_SUSTAINABILITY
Specifications
Specifications
250 mL
Spray Bottle