Learn More
MP Biomedicals™ RNase-Erase Decontaminants
Completely removes RNase contamination from glass and plastic surfaces
£153.00
Products 1
Product Code Brand Volume (Metric) Packaging Type Price Quantity & Availability  
Product Code Brand Volume (Metric) Packaging Type Price Quantity & Availability  
11421722
View Documents
MP Biomedicals™
04821682
250 mL Spray Bottle
£153.00
250mL
Estimated Shipment: 09-01-2025
to see stock.
Add to basket
Description
Description
Eliminates the need to bake glassware or use DEPC.
  • Leaves no residues that might inhibit enzymatic reactions
  • Supplied as ready-to-use formula