Learn More
Tocris Bioscience™ Ro 25-6981 maleate
GluN2B-selective NMDA antagonist
Brand:  Tocris Bioscience™ 1594/50
 View more versions of this product
Product Code. 18003882
  • £654.00 / 50mg
Explore more special offers
Description
Description
Ro 25-6981 maleate is a potent and selective activity-dependent blocker of NMDA receptors containing the GluN2B (formally NR2B) subunit. IC50 values are 0.009 and 52 μM for cloned receptor subunit combinations GluN1C/GluN2B and GluN1C/GluN2A respectively. Displays neuroprotectant effects in vivo and in vitro.
Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.
Specifications
Specifications
(αR,βS)-α-(4-Hydroxyphenyl)-β-methyl-4-(phenylmethyl)-1-piperidinepropanol maleate
>98%
Soluble to 100mM in DMSO
Observed MW: 455.55
1312991-76-6
50 mg
C22H29NO2.C4H4O4