Learn More
Tocris Bioscience™ Ro 25-6981 maleate
GluN2B-selective NMDA antagonist
£91.00 - £654.00
Specifications
Chemical Name or Material | (αR,βS)-α-(4-Hydroxyphenyl)-β-methyl-4-(phenylmethyl)-1-piperidinepropanol maleate |
---|---|
CAS | 1312991-76-6 |
Assay Percent Range | >98% |
Solubility | Soluble to 100mM in DMSO |
Molecular Formula | C22H29NO2.C4H4O4 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18795866
|
Tocris Bioscience™
1594/10 |
10 mg |
£175.00
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18003882
|
Tocris Bioscience™
1594/50 |
50 mg |
£654.00
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18014702
|
Tocris Bioscience™
1594/1 |
1 mg |
£91.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potent and selective activity-dependent blocker of NMDA receptors containing the GluN2B (formally NR2B) subunit. IC50 values are 0.009 and 52 μM for cloned receptor subunit combinations GluN1C/GluN2B and GluN1C/GluN2A respectively. Displays neuroprotectant effects in vivo and in vitro.Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.
Specifications
(αR,βS)-α-(4-Hydroxyphenyl)-β-methyl-4-(phenylmethyl)-1-piperidinepropanol maleate | |
>98% | |
C22H29NO2.C4H4O4 |
1312991-76-6 | |
Soluble to 100mM in DMSO | |
Observed MW: 455.55 |
Safety and Handling
Recommended Storage : Desiccate at Room Temperature
For Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.