Learn More
STEMCELL Technologies RoboSep™ Human Naïve B Cell Enrichment Kit with Filter Tips
EasySep™ kit with RoboSep™ buffer and RoboSep™ filter tips
Brand:  STEMCELL Technologies 19254RF
Encompass
Product Code. 17187911
  • £1173.00 / 1 set
Estimated Shipment: 23-10-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The EasySep™ Human Naïve B Cell Enrichment Kit is designed to isolate naïve B cells from peripheral blood mononuclear cells by negative selection
  • Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-naïve B cells and dextran-coated magnetic particles
  • The labeled cells are separated using the EasySep™ magnet without the use of columns
  • The desired cells are poured off into a new tube
Specifications
Specifications
B Cells
Cell Isolation
1 x 109 cells