missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Roll Form European Standard Pipe Markers with Die-cut Arrowheads - Air

Brand:  Brady™ 228244

Code : 50

Additional Details : Weight : 1.35000kg

 View more versions of this product

Product Code. 12890720

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

European Directives 67/548 and 88/379
Permanent Acrylic
Gloss
Roll Form Pipe Marker
Used for pipe markers and safety signs
220/Pk.
0.099 mm
100 mm
13 mm
Roll form Pipe Markers with liner, without pictograms - Air
White, Blue
Polyester
Sterile Air
English
-40°C to +110°C
150 x 100 mm
150.00 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers