missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Rose Bengal

£24.62 - £265.32

Chemical Identifiers

CAS 632-69-9
Molecular Formula C20H4Cl4I4Na2O5
Molecular Weight (g/mol) 1019.644
MDL Number MFCD00151169
InChI Key DPBQSRZQYAYDGY-UHFFFAOYSA-N
Synonym rose bengal disodium salt, r105 sodium, rose bengal sodium, rose-bengal 131 i natrium, food red no. 105, sodium salt, food red color no. 105, sodium salt, rosa bengala sodica 131 i inn-spanish, rose bengale 131 i sodique inn-french, sel disodique de rose bengale iodee 131 i, roseum bengalense 131 i natricum inn-latin
PubChem CID 87244310
IUPAC Name sodium;4,5,6,7-tetrachloro-3',6'-dihydroxy-2',4',5',7'-tetraiodospiro[2-benzofuran-3,9'-xanthene]-1-one
SMILES C1=C2C(=C(C(=C1I)O)I)OC3=C(C(=C(C=C3C24C5=C(C(=C(C(=C5Cl)Cl)Cl)Cl)C(=O)O4)I)O)I.[Na].[Na]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11428916
View Documents Promotion Details
Thermo Scientific Chemicals
A17053.14
25 g
£24.62
25g
Please to purchase this item. Need a web account? Register with us today!
11438916
View Documents
Thermo Scientific Chemicals
A17053.22
100 g
£70.25
100g
Please to purchase this item. Need a web account? Register with us today!
11448916
View Documents
Thermo Scientific Chemicals
A17053.36
500 g
£265.32
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Rose Bengal is a stain used in the eye drops to identify the eye damage. It finds application in the preparation of foraminifera for microscopic analysis, photosensitizing reactions and as a dye. In synthetic chemistry, it is utilized to convert triplet oxygen to singlet oxygen, which is used in [2+2] cycloadditions with alkenes. Its derivatives have important medical functions and used for the treatment of cancer. It acts as an alternative to phloxine B in Kreyberg's stain for keratin and mucus. As a fluorescent probe, it is used for the determination of NAD binding sites of alcohol dehydrogenase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

632-69-9
1019.644
DPBQSRZQYAYDGY-UHFFFAOYSA-N
87244310
C1=C2C(=C(C(=C1I)O)I)OC3=C(C(=C(C=C3C24C5=C(C(=C(C(=C5Cl)Cl)Cl)Cl)C(=O)O4)I)O)I.[Na].[Na]
C20H4Cl4I4Na2O5
MFCD00151169
rose bengal disodium salt, r105 sodium, rose bengal sodium, rose-bengal 131 i natrium, food red no. 105, sodium salt, food red color no. 105, sodium salt, rosa bengala sodica 131 i inn-spanish, rose bengale 131 i sodique inn-french, sel disodique de rose bengale iodee 131 i, roseum bengalense 131 i natricum inn-latin
sodium;4,5,6,7-tetrachloro-3',6'-dihydroxy-2',4',5',7'-tetraiodospiro[2-benzofuran-3,9'-xanthene]-1-one

Specifications

632-69-9
C20H4Cl4I4Na2O5
3645857
14,8262
Soluble in water.
C1=C2C(=C(C(=C1I)O)I)OC3=C(C(=C(C=C3C24C5=C(C(=C(C(=C5Cl)Cl)Cl)Cl)C(=O)O4)I)O)I.[Na].[Na]
1019.644
1017.65
>300°C
MFCD00151169
Light sensitive
rose bengal disodium salt, r105 sodium, rose bengal sodium, rose-bengal 131 i natrium, food red no. 105, sodium salt, food red color no. 105, sodium salt, rosa bengala sodica 131 i inn-spanish, rose bengale 131 i sodique inn-french, sel disodique de rose bengale iodee 131 i, roseum bengalense 131 i natricum inn-latin
DPBQSRZQYAYDGY-UHFFFAOYSA-N
sodium;4,5,6,7-tetrachloro-3',6'-dihydroxy-2',4',5',7'-tetraiodospiro[2-benzofuran-3,9'-xanthene]-1-one
87244310
Rose Bengal
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P362-P312-P321-P405-P403+P233-P501a

Warning

missing translation for 'einecsNumber' : 211-183-3

missing translation for 'rtecsNumber' : LM5920000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Rose Bengal

Signal Word
  • Warning

Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only