Learn More
BUCHI Rotavapor R-300 Vertical Condenser Glass Assembly
Vertical Condenser Glass Assembly for Rotavapor R-300, 1500 cm2. Assembly features a large top inlet for easy cleaning, connection for vapor temperature sensor, and inner glass tubing/plate. Includes a 1 liter receiving flask, tubing, and a ball joint clamp.
Brand:  BUCHI 11062432
 View more versions of this product
Product Code. 15530580
  • £1305.00 / Each
Estimated Shipment: 20-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Includes receiving flask (1 L), tubing, and a ball joint clamp
  • Does NOT include evaporating flask, vacuum seal, vapor duct, and condenser holder
  • For use with Rotavapor R-300, R-2XX
Specifications
Specifications
Borosilicate Glass (3.3)
Rotavap™ R-300, R-210, and R-215
No Safety Coating
Vertical Condensor
Receiving Flask, Tubing, and a Ball Joint Clamp
1 L
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BUCHI Rotavapor R-300 Vertical Condenser Glass Assembly > No Safety Coating