Learn More
Eppendorf™ Rotor Adapters for Centrifuge 5427 R
For 1.5/2mL tubes and cryotubes
£37.20 - £38.10
Specifications
Type Rotor Adapter
For Use With (Equipment) Rotors FA-45-12-17, FA-45-16-17 and FA-45-20-17
No. per Pack Set of 4
View More Specs
Products 2
Product Code Brand Capacity Price Quantity & Availability  
Product Code Brand Capacity Price Quantity & Availability  
13216619
View Documents
Eppendorf™
5820768002
1 x 1.5/2 mL Vessel
£37.20
Pack of 10
13226619
View Documents
Eppendorf™
5820769009
1 Cryotube
£38.10
Pack of 4
Description
Description
  • Rotors FA-45-12-17, FA-45-16-17 and FA-45-20-17
Specifications
Specifications
Rotor Adapter
Rotors FA-45-12-17, FA-45-16-17 and FA-45-20-17
Set of 4