Learn More
Eppendorf™ Rotor Lids
Designed for use with Eppendorf Rotors
Brand: Eppendorf™ 5427720006
147.00 GBP valid until 2024-12-31
Use promo code "23292" to get your promotional price.
Code : 99
Additional Details : Weight : 0.15700kg
Description
- Replacement rotor lids
- Aerosol-tight and aluminum lids available
- Available in a variety of sizes
Specifications
Rotor Lid | |
Aluminum | |
F–45-64-5-PCR | |
Non-aerosol-tight |