Learn More
IKA™ Round Bottom Flask Inlay
Round bottom flask inlay constructed of anodized aluminum
Brand: IKA™ 0025000638
This item is not returnable.
View return policy
Description
- Round bottom
- Anodized aluminum construction
- Temperature resistant
Specifications
50 mL | |
Round Bottom | |
180°C | |
Flask Carrier |