Learn More
Cowie Technology™ Round-Bottom Flask, PTFE
COWIE TECHNOLOGY™ Round-Bottom Flask, PTFE has a smooth internal finish and is usable at high temperatures.
Marke: Cowie Technology™ 040.010.19
Additional Details : Weight : 0.10000kg
Beschreibung
- Round Bottom Flasks have a smooth internal finish.
- PTFE used in the manufacture of Cowie™ PTFE Laboratory Products is pure, virgin material that conforms to FDA and USP Class VI requirements.
- Inert and usable to 280°C.
- Tapers match standard ground glass joints.
- Without sealing rings.
Spezifikation
32 mm | |
10 mL | |
19/26 | |
Up to 280°C |