missing translation for 'onlineSavingsMsg'
Learn More

Cowie Technology™ Round-Bottom Flask, PTFE

COWIE TECHNOLOGY™ Round-Bottom Flask, PTFE has a smooth internal finish and is usable at high temperatures.

£19.12 - £79.12

Specifications

Temperature Range (Metric) Up to 280°C
View More Specs

Products 6
Product Code Brand Capacity (Metric) Socket Joint Price Quantity & Availability  
Product Code Brand Capacity (Metric) Socket Joint Price Quantity & Availability  
10744523
View Documents Promotion Details
Cowie Technology™
040.025.19
25 mL 19/26 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10426982
View Documents Promotion Details
Cowie Technology™
040.010.19
10 mL 19/26 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10221572
View Documents Promotion Details
Cowie Technology™
040.025.24
25 mL 24/29 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10487552
View Documents Promotion Details
Cowie Technology™
040.050.29
50 mL 29/32 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10784903
View Documents Promotion Details
Cowie Technology™
040.050.24
50 mL 24/29 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10794903
View Documents Promotion Details
Cowie Technology™
040.100.29
100 mL 29/32 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

  • Round Bottom Flasks have a smooth internal finish.
  • PTFE used in the manufacture of Cowie™ PTFE Laboratory Products is pure, virgin material that conforms to FDA and USP Class VI requirements.
  • Inert and usable to 280°C.
  • Tapers match standard ground glass joints.
  • Without sealing rings.
Specifications

Specifications

Up to 280°C
Videos
SDS
Documents

Documents

Product Certifications