missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCAT Round Canisters
31.23 GBP valid until 2024-12-31
Use promo code "23290" to get your promotional price.
Use promo code "23290" to get your promotional price.
Click to view available options
Capacity (Metric):
2 L
4 L
5 L
10 L
25 L
50 L
60 L
Closure Size:
53B
83B
S65
Description
- Made of either robust HDPE or PP and therefore resistant to all solvents commonly used in the laboratory
- Available in the common threads S65, S70/71 and B83
- Filling capacities range from very compact 2 L to 4 L, 10 L, 25 L, 50 L and 60 L for the collection of voluminous solvents
Many individual features such as carrying handles, handles or measuring scales, which ensure safe handling of the hazardous substances
SCAT round canisters are a perfect alternative or supplement to conventional SCAT canisters to provide physical separation of solvents and prevent mixing of hazardous liquids.
Specifications
Capacity (English) | 0.5 gal. |
Capacity (Metric) | 2 L |
Closure Size | 53B |
Color | White (Translucent) |
Description | Round canister 2 L, B53, PE-HD, dimensions (W x H x D): 119 x 260 x 119 mm |
Dimensions (L x W x H) | 119 x 260 x 119 mm |
Disposable | Reusable |
Material | HDPE |
Mouth Style | B53 Thread |
Product Type | Canister |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'