Learn More
Cole-Parmer™ Round-Tapered Tip Tweezers
A slightly larger tip provides more surface area and gripping ability
Brand:  Cole-Parmer™ 07289-02
Product Code. 11715499
  • £74.85 / Each
Estimated Shipment: 21-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Finely crafted stainless steel tweezers
  • Feature hand finished points for accuracy and excellent tip hardness
  • Anti-glare finish reduces eye strain
  • Anti-magnetic, anti-acid and acid resistant
Specifications
Specifications
Round, Tapered
Carbon Fiber
Tweezers
Sterile
5 in.
12.7 cm
Soft and Strong Tweezers
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cole-Parmer™ Round-Tapered Tip Tweezers > Round-Tapered Tip Tweezers