missing translation for 'onlineSavingsMsg'
Learn More

Cytiva rProtein A Sepharose™ Fast Flow Affinity Media

Designed for purification of monoclonal and polyclonal antibodies at labs and process scales

Brand:  Cytiva 17-1279-03


 View more versions of this product

Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 10117549

  • £4863.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Composed of recombinant protein A coupled with cross-linked 4% agarose base matrix
  • High dynamic binding capacity, single-point attachment of protein A, with high recovery, high purity and low ligand leakage
  • No mammalian components involved during manufacturing process
  • Matrix: 4% cross-linked agarose
  • Average particle size: 90μm (d50V)
  • Ligand: 6mg rProtein A/mL medium (E. coli)
  • Binding capacity: ∽50mg human IgG/mL media
  • pH stability working range: 3 to 10
  • Pressure/Flow specification: base matrix 150 to 250cm/hr., 100kPa, XK 50/60 column, bed
TRUSTED_SUSTAINABILITY
Specifications

Specifications

200 mL
For PurifiCatIon of Monoclonal and Polyclonal Antibodies at Labs and Process Scales
50 mg Human lgG/mL medium2
150 to 250 cm/Hoμr
9.84 in.
100 kPa
2 to 9
Recombinant Protein A (E. coli)
Stable in aqueous Buffers commonly used in Protein A Chromatography
2°C to 8°C
rProtein A Sepharose Fast Flow Media
60 to 165 μm
25 cm
4% Cross-Linked Agarose
20% Ethanol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • rProtein A Sepharose Fast Flow

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.