missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Restek™ Rtx™-CLPesticides Capillary Columns
Designed to overcome the coelutions and analyte breakdown typically encountered in organochlorine pesticide and herbicide analyses
Brand: Restek™ 11123
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
This item is not returnable.
View return policy
Description
- Application-specific columns exceeding performance criteria for U.S. EPA methods 8081, 608, CLP
- Low bleed—ideal for GC/ECD or GC/MS analyses
- Baseline separations in less than 25 minutes
- Stable to 340°C
Specifications
350°C | |
Rtx™-CLP | |
0.25 μm | |
0.25 μm | |
Stainless Steel |
0.25 mm | |
Organochlorine Pesticides Analysis, Organochlorine Pesticides, Herbicides Analysis | |
-60°C to +320/340°C | |
30 m | |
Fused Silica |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction