Learn More
Abeba™ Rubber 5100 Shoes
ESD dissipative shoe is acid and base resistant.
Brand:  Abeba™ 5100/47
Additional Details : Weight : 0.95000kg
 View more versions of this product
Product Code. 15601828
  • £59.25 / 1 pair
Estimated Shipment: 08-08-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB A, FO SRC
  • Smooth leather white
  • PU midsole
  • Slip resistance special rubber outsole sole
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap non-foldable, adjustable
  • Leather top layer
  • Adjustable instep area
  • Lining microfiber, respiration absorbing
Specifications
Specifications
CE EN ISO 20347:2012 OB A, FO SRC
Microfiber
47
Rubber
White
Leather
Slip-resistant
5100