Learn More
Abeba™ Rubber 5150 Shoes
ESD dissipative shoe is acid and base resistant.
Brand: Abeba™ 5150/43
Additional Details : Weight : 0.95000kg
Description
- CE EN ISO 20347:2012 OB A, FO SRC
- Smooth leather
- Dark blue
- PU midsole
- Slip resistance special rubber outsole sole
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Heel strap non-foldable, adjustable
- Leather top layer
- Adjustable instep area
- Lining microfiber, respiration absorbing
Specifications
CE EN ISO 20347:2012 OB A, FO SRC | |
Microfiber | |
43 | |
Rubber |
Blue | |
Leather | |
Slip-resistant | |
5150 |