Learn More
Abeba™ Rubber 5310 Shoes
ESD dissipative shoe is acid and base resistant.
Brand:  Abeba™ 5310/38
Additional Details : Weight : 0.95000kg
 View more versions of this product
Product Code. 15671898
  • £56.49 / 1 pair
Estimated Shipment: 08-08-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB A, FO SRC
  • Smooth leather, perforated
  • Black
  • PU midsole
  • Slip resistance special rubber outsole sole
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap foldable, adjustable
  • Leather top layer
Specifications
Specifications
CE EN ISO 20347:2012 OB A, FO SRC
Leather
Slip-resistant
5310
Black
38
Rubber