Learn More
Abeba™ Rubber 9300 Shoes
Acid and base resistant. Rubber sole provides high slip resistance.
Brand:  Abeba™ 9300/36
Additional Details : Weight : 0.95000kg
 View more versions of this product
Product Code. 15623228
  • £54.30 / 1 pair
Estimated Shipment: 15-08-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB A, FO SRC
  • Smooth leather, perforated
  • White
  • PU midsole
  • Slip resistance special rubber outsole sole
  • Sanitized treated
  • Antistatic
  • Heel strap foldable, adjustable
  • Leather insole
Specifications
Specifications
CE EN ISO 20347:2012 OB A, FO SRC/li
Leather
Slip-resistant
9300
White
36
Rubber