Learn More
Rubidium hydroxide, 50% w/w aq. soln., 99.6+% (metals basis), Thermo Scientific Chemicals
£113.57 - £553.76
Chemical Identifiers
CAS | 1310-82-3 |
---|---|
Molecular Formula | HORb |
Molecular Weight (g/mol) | 102.48 |
MDL Number | MFCD00011194 |
InChI Key | CPRMKOQKXYSDML-UHFFFAOYSA-M |
Synonym | rubidium hydroxide, rubidium hydroxide rb oh, rboh, rubidium 1+ hydroxide, rubidium hydroxide solution, rubidiumol, rubidium hydoxide, rubidium hydroxide sup 81 rb, rubidium monohydroxide, rb.hoShow More |
PubChem CID | 62393 |
ChEBI | CHEBI:32108 |
IUPAC Name | rubidium(1+);hydroxide |
SMILES | [OH-].[Rb+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11335799
|
Thermo Scientific Alfa Aesar
010565.NC |
10 g |
£113.57
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11345799
|
Thermo Scientific Alfa Aesar
010565.NE |
50 g |
£553.76
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Rubidium Hydroxide is used to give fireworks a violet color. It is a notable ntermediate in the synthesis of other rubidium compounds. It is used in photography and storage batteries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1310-82-3 | |
102.48 | |
CPRMKOQKXYSDML-UHFFFAOYSA-M | |
62393 | |
rubidium(1+);hydroxide |
HORb | |
MFCD00011194 | |
rubidium hydroxide, rubidium hydroxide rb oh, rboh, rubidium 1+ hydroxide, rubidium hydroxide solution, rubidiumol, rubidium hydoxide, rubidium hydroxide sup 81 rb, rubidium monohydroxide, rb.hoShow More | |
CHEBI:32108 | |
[OH-].[Rb+] |
Specifications
1310-82-3 | |
Liquid | |
RbOH | |
UN2677 | |
rubidium hydroxide, rubidium hydroxide rb oh, rboh, rubidium 1+ hydroxide, rubidium hydroxide solution, rubidiumol, rubidium hydoxide, rubidium hydroxide sup 81 rb, rubidium monohydroxide, rb.ho | |
CPRMKOQKXYSDML-UHFFFAOYSA-M | |
rubidium(1+);hydroxide | |
62393 | |
102.48 (Anhydrous) | |
50% w/w aq. soln. | |
Rubidium hydroxide |
Yellow | |
HORb | |
MFCD00011194 | |
14,8285 | |
Soluble in ethanol and water. | |
[OH-].[Rb+] | |
102.48 | |
CHEBI:32108 | |
≥99.6% | |
(metals basis) |
Safety and Handling
GHS H Statement
H314-H302
Causes severe skin burns and eye damage.
Harmful if swallowed.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: RUBIDIUM HYDROXIDE SOLUTION
missing translation for 'einecsNumber' : 215-186-0
missing translation for 'rtecsNumber' : VL8750000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.