Learn More
Rubidium sulfate, 99% (metals basis), Thermo Scientific Chemicals
Description
Rubidium sulfate is used in hematology stain. It is also used as the raw materials for making kinds of Rubidium salts. It is fused on a ceramic bead and used as thermoionic detector. It is also used as a cathartic.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 1050°C |
Physical Form | Crystalline |
Quantity | 25 g |
Synonym | rubidium sulfate, rubidium sulphate, dirubidium sulfate, rubidium sulfate rb2so4, sulfuric acid, dirubidium salt, rubidium sulfate rb2 so4, dirubidium 1+ sulfate, rubidiumsulfat, sulfuric acid, rubidium salt 1:2, rubidium 1+ sulfate |
Solubility Information | Soluble in water (424mg/ml at 10°C). |
Formula Weight | 267 |
Percent Purity | 99% |
Odor | Odorless |
Refractive Index | 1.513 |
Density | 3.613 g/mL |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
W06J054 | Certificate of Analysis | 21/09/2023 | 013493.22,013493.14,013493.36 |
X27H020 | Certificate of Analysis | 12/04/2022 | 013493.22,013493.14,013493.36 |
D21K13 | Certificate of Analysis | 08/12/2021 | 013493.22,013493.14,013493.36 |
M15F014 | Certificate of Analysis | 08/12/2021 | 013493.22,013493.14,013493.36 |
G11N07 | Certificate of Analysis | 08/12/2021 | 013493.22,013493.14,013493.36 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.