missing translation for 'onlineSavingsMsg'
Learn More
Rubidium sulfate, 99% (metals basis), Thermo Scientific Chemicals Product Code.: 11450320

Rubidium sulfate, 99% (metals basis), Thermo Scientific Chemicals

Product Code. 11450320
500 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11450320

Brand: Thermo Scientific Alfa Aesar 013493.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rubidium sulfate is used in hematology stain. It is also used as the raw materials for making kinds of Rubidium salts. It is fused on a ceramic bead and used as thermoionic detector. It is also used as a cathartic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7488-54-2
Molecular Formula H2O4RbS
Molecular Weight (g/mol) 183.54
MDL Number MFCD00011190
InChI Key XNNUAZGCOYTNQE-UHFFFAOYSA-N
PubChem CID 197088
IUPAC Name sulfuric acid rubidium
SMILES [Rb].OS(O)(=O)=O

Specifications

Melting Point 1050°C
Physical Form Crystalline
Quantity 500 g
Synonym rubidium sulfate, rubidium sulphate, dirubidium sulfate, rubidium sulfate rb2so4, sulfuric acid, dirubidium salt, rubidium sulfate rb2 so4, dirubidium 1+ sulfate, rubidiumsulfat, sulfuric acid, rubidium salt 1:2, rubidium 1+ sulfate
Solubility Information Soluble in water (424mg/ml at 10°C).
Formula Weight 267
Percent Purity 99%
Odor Odorless
Refractive Index 1.513
Density 3.613 g/mL
Assay (metals basis)
Chemical Name or Material Rubidium sulfate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW06J054 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog Number013493.22,013493.14,013493.36
Lot NumberR12J008 Certificate TypeCertificate of Analysis Date22/05/2023 Catalog Number013493.36
Lot NumberP13J002 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog Number013493.36
Lot NumberX27H020 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number013493.22,013493.14,013493.36
Lot NumberD21K13 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number013493.22,013493.14,013493.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Rubidium sulfate, 99% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.